Learn More
Biomatik Corporation Bovine Peroxisome Proliferator Activated Receptor Gamma (PPARg) ELISA Kit
Supplier:  Biomatik Corporation EKU06564
Encompass_Preferred
Bovine Peroxisome Proliferator Activated Receptor Gamma (PPARg) ELISA Kit for detecting PPARg in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.133ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O18971. Gene ID: 281993. Target Synonyms: PPAR-G; PPARG1; PPARG2; NR1C3; Glitazone Receptor; Nuclear Receptor Subfamily 1 Group C Member 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4358