Learn More
Lonza Walkersville Bovine Pituitary Extract (BPE), 13mg/mL, 2 mL
Catalog No. NC9272093
Encompass
Bovine Pituitary Extract is useful in the culture of various epithelial cell types. The product is included in SingleQuots Kits associated with various media systems for epithelial cell types. The product is available at a concentration of 13 mg/ml.Content:1 x Vial Containing 13 mg/mL BPE 2 mL
Catalog No. NC9272093 Supplier Lonza Walkersville Supplier No. CC4009
Bovine Pituitary Extract is useful in the culture of various epithelial cell types. The product is included in SingleQuots Kits associated with various media systems for epithelial cell types. The product is available at a concentration of 13 mg/ml.Content:1 x Vial Containing 13 mg/mL BPE 2 mL