Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bovine Pituitary Extract (BPE)

Catalog No. 13028014
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-028-014 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-028-014 Supplier Gibco™ Supplier No. 13028014
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Bovine Pituitary Extract (BPE)

The protein concentration of each lot of Bovine Pituitary Extract is determined by the Bradford method. Each vial of BPE contains not less than 25 mg of protein.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Cell Type Mammalian Cells
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Bovine Pituitary Extract
pH 6 to 8
Quantity 25 mg
Shipping Condition Dry Ice
Are the Keratinocyte SFM basal medium and supplement kit available separately?

The basal medium is not available for purchase individually, and can only be ordered as part of the Keratinocyte SFM kit (catalog # 17005-042). The supplements are available separately: Bovine Pituitary Extract is catalog # 13028-014 and the EGF is catalog # PHG0311.

Which serum-free media can be used to culture mouse keratinocytes?

The Keratinocyte SFM without calcium chloride (Cat. No. 37010-022) is appropriate for the culture of mouse keratinocytes. The medium should be supplemented with calcium chloride at a final concentration of 0.02 mM.

Which serum-free media can be used to culture human keratinocytes?

There are a variety of options for serum-free culture of human keratinocytes. Keratinocyte SFM (Cat. No. 17005-042) is a widely used kit that includes the basal medium, bovine pituitary extract (BPE) and Epidermal Growth Factor (EGF). If you would like a serum-free, defined culture system, the options are the Defined Keratinocyte SFM kit (Cat. No. 10744-019) or Epilife medium (Cat. No. M-EPI-500-CA) supplemented with EDGS (Cat. No. S-012-5). If you would like an animal origin free defined system, Epilife medium (Cat. No. M-EPI-500-CA) with S7 supplement (Cat. No. S-017-5) is an excellent choice.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.