Learn More
Sciencell Research Laboratories BOVINE PLASMA FIBRONECTIN 1MG
Catalog No. NC0009247
Encompass
Fibronectin is a 440-500 kDa glycoprotein found both as cell surface proteins and in plasma. BPF is purified from bovine plasma with affinity chromatography, serving as an ideal substrate to support cell adhesion and spreading.
Catalog No. NC0009247 Supplier Sciencell Research Laboratories Supplier No. 8248
Fibronectin is a 440-500 kDa glycoprotein found both as cell surface proteins and in plasma. BPF is purified from bovine plasma with affinity chromatography, serving as an ideal substrate to support cell adhesion and spreading.