Learn More
Biomatik Corporation Bovine Platelet Derived Growth Factor Subunit B (PDGFB) ELISA Kit
Supplier:  Biomatik Corporation EKU11344
Encompass_Preferred
Bovine Platelet Derived Growth Factor Subunit B (PDGFB) ELISA Kit for detecting PDGFB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B1H0W5. Gene ID: 540106. Target Synonyms: PDGF-B; PDGF2; SIS; SSV; C-sis; Simian Sarcoma Viral (v-sis) Oncogene Homolog; Platelet Derived Growth Factor Beta Polypeptide; Proto-oncogene c-Sis; Becaplermin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8163