Learn More
Biomatik Corporation Bovine Pleiotrophin (PTN) CLIA Kit
Supplier:  Biomatik Corporation EKU09660
Encompass_Preferred
Bovine Pleiotrophin (PTN) CLIA Kit for detecting PTN in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.15pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P21782. Gene ID: 280904. Target Synonyms: HARP; HBGF8; HBNF; NEGF1; Heparin Binding Growth Factor 8; Neurite Growth-Promoting Factor 1; Heparin Affin Regulatory Peptide; Heparin Binding Growth Associated Molecule. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7259