Learn More
Biomatik Corporation Bovine Polymeric Immunoglobulin Receptor (PIGR) ELISA Kit
Supplier:  Biomatik Corporation EKU06741
Encompass_Preferred
Bovine Polymeric Immunoglobulin Receptor (PIGR) ELISA Kit for detecting PIGR in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.238ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P81265. Gene ID: 281401. Target Synonyms: SC; Secretory Component; Hepatocellular carcinoma-associated protein TB6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7548