Learn More
American Research Products Inc BoVine Pregnancy Specific Prot
Catalog No. 5000329849
Encompass
BoVine Pregnancy Specific Protein B (PSPB) ELISA Kit from CUSABIO Detection Range 0 06 ng/mL-24 ng/mL Sensitivity 0 06 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29849 Supplier American Research Products Inc Supplier No. CSBE13353B
BoVine Pregnancy Specific Protein B (PSPB) ELISA Kit from CUSABIO Detection Range 0 06 ng/mL-24 ng/mL Sensitivity 0 06 ng/mL Sample serum plasma tissue homogenates Method Competitive