Learn More
Biomatik Corporation Bovine Prion Protein (PRNP) ELISA Kit
Supplier:  Biomatik Corporation EKU12180
Encompass_Preferred
Bovine Prion Protein (PRNP) ELISA Kit for detecting PRNP in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10279. Gene ID: 281427. Target Synonyms: CD230; GSS; ASCR; CJD; PRIP; PrP; PrP2730; PrPc; Major prion protein; Creutzfeldt-Jakob Disease; Gerstmann-Strausler-Scheinker Syndrome; Fatal Familial Insomnia. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4947