Learn More
American Research Products Inc BoVine procalcitonin PCT ELI
Catalog No. 5000330074
Encompass
BoVine procalcitonin (PCT) ELISA kit from CUSABIO Detection Range 39 06 pg/mL-2500 pg/mL Sensitivity 9 77 pg/mL Sample serum plasma tissue Homogenates Method Sandwich
Catalog No. 50-003-30074 Supplier American Research Products Inc Supplier No. CSBE15004B
BoVine procalcitonin (PCT) ELISA kit from CUSABIO Detection Range 39 06 pg/mL-2500 pg/mL Sensitivity 9 77 pg/mL Sample serum plasma tissue Homogenates Method Sandwich