Learn More
Biomatik Corporation Bovine Prothymosin Alpha (PTMa) ELISA Kit
Supplier:  Biomatik Corporation EKU12339
Encompass_Preferred
Bovine Prothymosin Alpha (PTMa) ELISA Kit for detecting PTMa in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01252. Gene ID: 786336. Target Synonyms: TMSA; PTM-A; Pro-Thymosin A; Gene Sequence 28; Thymosin alpha-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7980