Learn More
American Research Products Inc Bovine PTH ELISA Kit
Catalog No. 5000458638
Encompass
This Bovine Parathyroid Hormone (PTH) ELISA Kit is a precoated Competitive Inhibition ELISA assay designed to detect and quantify the target analyte in serum plasma tissue homogenates cell lysates cell culture supernatants and other biological fluids This product is for Research Use Only
Catalog No. 50-004-58638 Supplier American Research Products Inc Supplier No. RDRPTHB
This Bovine Parathyroid Hormone (PTH) ELISA Kit is a precoated Competitive Inhibition ELISA assay designed to detect and quantify the target analyte in serum plasma tissue homogenates cell lysates cell culture supernatants and other biological fluids This product is for Research Use Only