Learn More
American Research Products Inc BoVine pulmonary surfactant as
Catalog No. 5000327471
Encompass
BoVine pulmonary surfactant-associated protein B SP-B ELISA Kit from CUSABIO Detection Range 9 37 ng/mL-600 ng/mL Sensitivity 2 34 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27471 Supplier American Research Products Inc Supplier No. CSBE13834B
BoVine pulmonary surfactant-associated protein B SP-B ELISA Kit from CUSABIO Detection Range 9 37 ng/mL-600 ng/mL Sensitivity 2 34 ng/mL Sample serum plasma tissue homogenates Method Sandwich