Learn More
American Research Products Inc BoVine pulmonary surfactant as
Supplier:  American Research Products Inc CSBE13835B
Encompass
BoVine pulmonary surfactant-associated protein C SP-C ELISA Kit from CUSABIO Detection Range 12 5 ng/mL-800 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29291