Learn More
Biomatik Corporation Bovine Reactive Oxygen Species Modulator 1 (ROMO1) ELISA Kit
Supplier:  Biomatik Corporation EKU11389
Encompass_Preferred
Bovine Reactive Oxygen Species Modulator 1 (ROMO1) ELISA Kit for detecting ROMO1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3SZV8. Gene ID: 618530. Target Synonyms: C20orf52; MTGMP; Glyrichin; Mitochondrial Targeting GXXXG Protein; Epididymis tissue protein Li 175; Protein MGR2 homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8152