Learn More
Biomatik Corporation Bovine S100 Calcium Binding Protein A7 (S100A7) ELISA Kit
Supplier:  Biomatik Corporation EKU12192
Encompass_Preferred
Bovine S100 Calcium Binding Protein A7 (S100A7) ELISA Kit for detecting S100A7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q28050. Gene ID: 282344. Target Synonyms: S100-A7; PSOR1; S100A7c; Psoriasin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5956