Learn More
Biomatik Corporation Bovine S100 Calcium Binding Protein A8 (S100A8) ELISA Kit
Supplier:  Biomatik Corporation EKU07177
Encompass_Preferred
Bovine S100 Calcium Binding Protein A8 (S100A8) ELISA Kit for detecting S100A8 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 78.12-5000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28782. Gene ID: 616818. Target Synonyms: MRP8; MIF; 60B8AG; CAGA; CFAG; CGLA; CP-10; L1Ag; NIF; P8; Calgranulin A; Cystic fibrosis antigen; Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7510