Learn More
Biomatik Corporation Bovine Secretory Immunoglobulin A (sIgA) CLIA Kit
Supplier:  Biomatik Corporation EKU09842
Encompass_Preferred
Bovine Secretory Immunoglobulin A (sIgA) CLIA Kit for detecting sIgA in serum, plasma, saliva, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 68.6-50,000pg/ml. Sensitivity: 29.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Target Synonyms: S-IgA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6273