Learn More
American Research Products Inc BoVine Selenoprotein P SEPP1
Catalog No. 5000327669
Encompass
BoVine Selenoprotein P (SEPP1) ELISA kit from CUSABIO Detection Range 10 ng/mL-160 ng/mL Sensitivity 5 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27669 Supplier American Research Products Inc Supplier No. CSBEL021018BO
BoVine Selenoprotein P (SEPP1) ELISA kit from CUSABIO Detection Range 10 ng/mL-160 ng/mL Sensitivity 5 ng/mL Sample serum plasma tissue homogenates Method Competitive