Learn More
Boston Bioproducts Inc Bovine Serum Albumin, BSA (5%, TBS) - 250ml
Catalog No. NC2961858
Encompass
The 5% Bovine Serum Albumin (BSA) Buffer is used in laboratory settings to stabilize enzymes during reactions, enhance signals in immunoassays through non-specific binding, and as a protein standard. BSA is used as a diluent of primary and secondary antibodies for immunoassays and as a blocking agent. This product is buffered with Tris Buffered Saline.
Catalog No. NC2961858 Supplier Boston Bioproducts Inc Supplier No. IBB187250ML
The 5% Bovine Serum Albumin (BSA) Buffer is used in laboratory settings to stabilize enzymes during reactions, enhance signals in immunoassays through non-specific binding, and as a protein standard. BSA is used as a diluent of primary and secondary antibodies for immunoassays and as a blocking agent. This product is buffered with Tris Buffered Saline.