Learn More
AG Scientific Inc Bovine Serum Albumin, 100 G
Supplier:  AG Scientific Inc B3371100GM
Encompass_Preferred
Bovine serum albumin (BSA) is a high-purity albumin protein derived from the blood plasma of cows. Among its most common research uses is the culturing of mammalian cells, an essential function for many biochemical, pharmacological and biological studies. In addition to its use as a culturing agent, significant research has been conducted into BSA itself. Of particular importance has been inquiry into BSA present in cow's milk as a possible causative agent of insulin-dependent diabetes mellitus. Nanoparticles of albumin have been explored as drug delivery systems due to their biocompatible nature. BSA can also be used as a diluting component in vaccines.CAS Number: 9048-46-8Molecular Weight: ~66 kDaSolubility: Water: clear to slightly hazy faint yellow to yellow green solutionStorage Temperature: +2-8CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6847