Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin | 9048-46-8 | MFCD00130384 | 10 g
Catalog No. NC1069537
Encompass
Bovine Serum Albumin | Purity: 96% | Mol Wt: ~66 kDa | 9048-46-8 | MFCD00130384 | 10 g
Catalog No. NC1069537 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A788810G
Bovine Serum Albumin | Purity: 96% | Mol Wt: ~66 kDa | 9048-46-8 | MFCD00130384 | 10 g