Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin, 9048-46-8, MFCD00130384, 10 g
Catalog No. NC1498732
Encompass
Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis), Synonym: Albumin bovine serum, BSA, Bovine albumin. This is a cold alcohol fractionated albumin that is produced by Sigma-Aldrich with a stricter adherence to the original Cohn method of fractionation than other commercially available bovine albumins.
Catalog No. NC1498732 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A561110G
Add to Cart
Add to Cart
Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis), Synonym: Albumin bovine serum, BSA, Bovine albumin. This is a cold alcohol fractionated albumin that is produced by Sigma-Aldrich with a stricter adherence to the original Cohn method of fractionation than other commercially available bovine albumins.