Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin, 9048-46-8, MFCD00130384, 50g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A703050G
Encompass
Heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%, Synonym: Albumin bovine serum, BSA, Bovine albumin
Catalog No. 50-165-7300