Learn More
Medchemexpress LLC HY-D0842 1g Medchemexpress, Bovine Serum Albumin CAS:9048-46-8 Purity:>98%
SDP
Catalog No. 501933199
Encompass_Preferred
Medchemexpress, HY-D0842 1g Bovine Serum Albumin CAS:9048-46-8 Bovine Serum Albumin (BSA) is a 583-residue protein consisting of three homologous all-α domains, organized in a heart-shaped structure. BSA is a globular protein that is used in numerous biochemical applications. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3199 Supplier Medchemexpress LLC Supplier No. HYD08421G
Medchemexpress, HY-D0842 1g Bovine Serum Albumin CAS:9048-46-8 Bovine Serum Albumin (BSA) is a 583-residue protein consisting of three homologous all-α domains, organized in a heart-shaped structure. BSA is a globular protein that is used in numerous biochemical applications. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.