Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boston Bioproducts Inc Bovine Serum Albumin, 3% w/v, 500 ml, PBS

Supplier:  Boston Bioproducts Inc IBB101500ML

Encompass_Preferred

Bovine Serum Albumin, 3% w/v, 500 ml, PBS

Catalog No. NC3354784


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.