Learn More
Selleck Chemical LLC Bovine Serum Albumin P1245-250MG
SDP
Catalog No. 5000039798
Encompass_Preferred
Bovine serum albumin (BSA Albumin bovine serum) is an efficient passivating agent that inhibits nonspecific protein binding in assays such as ELISA It is used to coat hydrophobic substrates enhancing anti-fouling properties and reducing immune system interactions
Catalog No. 50-000-39798 Supplier Selleck Chemical LLC Supplier No. P1245250MG
Bovine serum albumin (BSA Albumin bovine serum) is an efficient passivating agent that inhibits nonspecific protein binding in assays such as ELISA It is used to coat hydrophobic substrates enhancing anti-fouling properties and reducing immune system interactions