Learn More
Boston Bioproducts Inc Bovine Serum Albumin, BSA (3%, TBST) - 500ml
Catalog No. NC2395533
Encompass
The 3% Bovine Serum Albumin (BSA) Buffer is used in laboratory settings to stabilize enzymes during reactions, enhance signals in immunoassays through non-specific binding, and as a protein standard. BSA is used as a diluent of primary and secondary antibodies for immunoassays and as a blocking agent. This product is buffered with Tris Buffered Saline - Tween.
Catalog No. NC2395533 Supplier Boston Bioproducts Inc Supplier No. IBB112500ML
The 3% Bovine Serum Albumin (BSA) Buffer is used in laboratory settings to stabilize enzymes during reactions, enhance signals in immunoassays through non-specific binding, and as a protein standard. BSA is used as a diluent of primary and secondary antibodies for immunoassays and as a blocking agent. This product is buffered with Tris Buffered Saline - Tween.