Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin | 9048-46-8 | MFCD00130384 | 50 g
Catalog No. NC1315305
Encompass
Bovine Serum Albumin | Purity: ≥96% | Mol Wt: ~66 kDa | 9048-46-8 | MFCD00130384 | 50 g
Catalog No. NC1315305 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A450350G
Bovine Serum Albumin | Purity: ≥96% | Mol Wt: ~66 kDa | 9048-46-8 | MFCD00130384 | 50 g