Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin, 9048-46-8, MFCD00130384, 50 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A964750G
Encompass
Heat shock fraction, pH 7, ≥98%, Synonym: Albumin bovine serum, BSA, Bovine albumin. Bovine Serum Albumin has been used as a supplement in the medium prepared for culturing of oligodendrocytes.
Catalog No. NC1628628