Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bovine serum albumin | 9048-46-8 | 98.0% | 66,430 Da | 10 MG
SDP

Catalog No. 5000297772
Encompass_Preferred

Bovine serum albumin (BSA) is a globular serum protein supplied as a light yellow powder. It is commonly used as a carrier protein, blocking agent in immunoassays, a standard in protein assays and electrophoresis, and as a stabilizer in biochemical and cell culture applications.

  • High purity: 98.0%.
  • Form: light yellow to yellow powder, supplied as a dry solid.
  • Common applications: carrier protein, blocking protein in immunoassays, cell culture supplement, standard in protein assays and electrophoresis.
  • Stabilizes proteins and helps prevent hydrolysis or denaturation.
  • Molecular weight: ~66.4 kDa (mature 583-residue form).
  • CAS number: 9048-46-8.

Catalog No. 50-002-97772 Supplier Medchemexpress LLC Supplier No. HYCP00210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bovine serum albumin (BSA) is a globular serum protein supplied as a light yellow powder. It is commonly used as a carrier protein, blocking agent in immunoassays, a standard in protein assays and electrophoresis, and as a stabilizer in biochemical and cell culture applications.

  • High purity: 98.0%.
  • Form: light yellow to yellow powder, supplied as a dry solid.
  • Common applications: carrier protein, blocking protein in immunoassays, cell culture supplement, standard in protein assays and electrophoresis.
  • Stabilizes proteins and helps prevent hydrolysis or denaturation.
  • Molecular weight: ~66.4 kDa (mature 583-residue form).
  • CAS number: 9048-46-8.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.