Learn More
Medchemexpress LLC Bovine Serum Albumin | 9048-46-8 | 98.2% | 500 MG
SDP
Catalog No. 5000411039
Encompass_Preferred
Bovine Serum Albumin (BSA) is a 583-residue protein consisting of three homologous all-α domains, organized in a heart-shaped structure. BSA is a globular protein that is used in numerous biochemical applications.
  • Consists of three homologous all-α domains
  • Organized in a heart-shaped structure
  • Globular protein for biochemical applications
  • Constitutes approximately 60% of all plasma protein
  • Binds and transports numerous physiological and non-physiological ligands
  • Contains 17 disulfide bonds and one unpaired cysteine (Cys34)
  • Benefits cell growth regardless of aggregated state
  • Confirms lack of cytotoxicity through membrane permeabilization assays
  • Increases viability of treated cells by 300-400%
Catalog No. 50-004-11039 Supplier Medchemexpress LLC Supplier No. HYD0842500MG
Bovine Serum Albumin (BSA) is a 583-residue protein consisting of three homologous all-α domains, organized in a heart-shaped structure. BSA is a globular protein that is used in numerous biochemical applications.
  • Consists of three homologous all-α domains
  • Organized in a heart-shaped structure
  • Globular protein for biochemical applications
  • Constitutes approximately 60% of all plasma protein
  • Binds and transports numerous physiological and non-physiological ligands
  • Contains 17 disulfide bonds and one unpaired cysteine (Cys34)
  • Benefits cell growth regardless of aggregated state
  • Confirms lack of cytotoxicity through membrane permeabilization assays
  • Increases viability of treated cells by 300-400%