Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bovine serum albumin-AF647 | >98% | 100 UL
SDP

Catalog No. 5000425720
Encompass_Preferred

Bovine Serum Albumin-AF647 (BSA-AF647) is a biological material and organic compound extensively utilized in life science research. This liquid formulation has a concentration of 1.0 mg/mL and a purity of >98%. It appears colorless to light yellow and is derived from bovine serum.

  • Colorless to light yellow liquid appearance
  • Derived from bovine serum
  • Concentration: 1.0 mg/mL
  • AF647/Protein ratio: ≈3.32
  • Formulated in 0.01M TBS (pH 7.4) with 0.03% Proclin300 and 50% Glycerol
  • Store at -20°C for 2 years, protect from light
  • Targeted for biochemical assay reagents
  • Widely used in life science research

Catalog No. 50-004-25720 Supplier Medchemexpress LLC Supplier No. HYNP048100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bovine Serum Albumin-AF647 (BSA-AF647) is a biological material and organic compound extensively utilized in life science research. This liquid formulation has a concentration of 1.0 mg/mL and a purity of >98%. It appears colorless to light yellow and is derived from bovine serum.

  • Colorless to light yellow liquid appearance
  • Derived from bovine serum
  • Concentration: 1.0 mg/mL
  • AF647/Protein ratio: ≈3.32
  • Formulated in 0.01M TBS (pH 7.4) with 0.03% Proclin300 and 50% Glycerol
  • Store at -20°C for 2 years, protect from light
  • Targeted for biochemical assay reagents
  • Widely used in life science research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.