Learn More
Medchemexpress LLC Bovine serum albumin (biotinylated) | 9048-46-8 | 1 MG
SDP
Catalog No. 5000425780
Encompass_Preferred
Bovine serum albumin (biotinylated) is a biotinylated bovine serum albumin that selectively binds to different vascular subsets in lymphoid tissue. It can be used as a histochemical probe and a targeting ligand for drug delivery.
  • Can be used as a reliable probe for the histological identification of select microvascular endothelia in lymphoid tissues.
  • Investigated as a targeting ligand in the design of nanoparticle-based delivery systems for anticancer drugs.
  • Serves as a platform that can be conjugated with the magnetic resonance imaging (MRI) contrast agent gadolinium diethylenetriamine pentaacetate (GdDTPA) to form a composite probe, enabling its application in studying fibroblast migration.
Catalog No. 50-004-25780 Supplier Medchemexpress LLC Supplier No. HYNP002D1MG
Bovine serum albumin (biotinylated) is a biotinylated bovine serum albumin that selectively binds to different vascular subsets in lymphoid tissue. It can be used as a histochemical probe and a targeting ligand for drug delivery.
  • Can be used as a reliable probe for the histological identification of select microvascular endothelia in lymphoid tissues.
  • Investigated as a targeting ligand in the design of nanoparticle-based delivery systems for anticancer drugs.
  • Serves as a platform that can be conjugated with the magnetic resonance imaging (MRI) contrast agent gadolinium diethylenetriamine pentaacetate (GdDTPA) to form a composite probe, enabling its application in studying fibroblast migration.