Learn More
Medchemexpress LLC Bovine Serum Albumin (Biotinylated) | 9048-46-8 | 5 MG
SDP
Catalog No. 5000425307
Encompass_Preferred
Bovine Serum Albumin (Biotinylated) is a biotinylated bovine serum albumin that selectively binds to different vascular subsets in lymphoid tissue. It is intended for research use only.
  • Can be used as a histochemical probe and a targeting ligand for drug delivery.
  • Reliable probe for histological identification of select microvascular endothelia in lymphoid tissues.
  • Investigated as a targeting ligand in the design of nanoparticle-based delivery systems for anticancer drugs.
  • Serves as a platform that can be conjugated with MRI contrast agents to study fibroblast migration.
Catalog No. 50-004-25307 Supplier Medchemexpress LLC Supplier No. HYNP002D5MG
Bovine Serum Albumin (Biotinylated) is a biotinylated bovine serum albumin that selectively binds to different vascular subsets in lymphoid tissue. It is intended for research use only.
  • Can be used as a histochemical probe and a targeting ligand for drug delivery.
  • Reliable probe for histological identification of select microvascular endothelia in lymphoid tissues.
  • Investigated as a targeting ligand in the design of nanoparticle-based delivery systems for anticancer drugs.
  • Serves as a platform that can be conjugated with MRI contrast agents to study fibroblast migration.