Learn More
Bioworld Bovine Serum Albumin, Fraction V - Fatty Acid Free 5g
Supplier:  Bioworld 220700231
Encompass_Preferred
Fatty acid-free BSA is a highly purified, lyophilized powder. This product is processed to provide a minimum of 98% albumin and to be essentially free of non-esterified fatty acids. The pH is adjusted to 7.0. It is used as a blocking reagent in EIA, RIA and ELISA systems, where serum fatty acids may interfere with the application. Specifications: Protein: ≥98.0%, Purity (albumin): ≥98.0%, Non esterified fatty acids: <0.01%, Heavy metals (pb): ≤10ppm, Ash: <2.0%, Bovine IgG: Not detected, Solubility (5% Water): Pass, pH (5%, water): 6 - 7.5, Ships at ambient. Appearance Form : powder, Appearance Color : Brown. 5g
Catalog No. 50-253-934