Learn More
Bioworld Bovine Serum Albumin (BSA), Fraction V - Fatty Acid Free, 10 g
Supplier:  Bioworld 220700232
Encompass_Preferred
Fatty acid-free BSA is a highly purified, lyophilized powder. This product is processed to provide a minimum of 98% albumin and to be essentially free of non-esterified fatty acids. The pH is adjusted to 7.0. It is used as a blocking reagent in EIA, RIA and ELISA systems, where serum fatty acids may interfere with the application.SpecificationsProtein: ≥98.0%Purity (albumin): ≥98.0%Non esterified fatty acids: <0.01%Heavy metals (pb): ≤10ppmAsh: <2.0%Bovine IgG: Not detectedSolubility (5% Water): PasspH (5%, water): 6 - 7.5Ships at ambient.
Catalog No. 50-253-935