Learn More
Bioworld Bovine Serum Albumin (BSA), Fraction V - Fine Powder, 1 kg
Supplier:  Bioworld 220700163
Encompass_Preferred
Standard grade BSA is a highly purified, lyophilized powder. Its characteristics include minimum 98% albumin, pH adjusted to 7.0, caprylate-free, and low conductivity. It is used in bacterial media,enzyme systems, and as a protein base.
Catalog No. 50-253-918