Learn More
Bioworld Bovine Serum Albumin (BSA), Fraction V - Low Electrolyte, 100 g
Supplier:  Bioworld 220700242
Encompass_Preferred
low-electrolyte BSA is a highly purified, lyophilized powder. This product is processed to provide a minimum of 97% albumin. The pH is adjusted to 5.2. Conductivity of < 1000 µS in a 10% aqueous solution is typical. It is used in media, enzyme systems, and as a stabilizer for EIA and RIA techniques, where high purity and low electrolyte levels are critical.SpecificationsShips at ambient.
Catalog No. 50-253-938