Learn More
Gold Biotechnology Inc Bovine Serum Albumin (BSA), Fraction V, Fatty Acid Free for tissue culture 1 kg
SDP
Catalog No. NC2112210
Encompass_Preferred
GoldBio's Reagent Grade, Fatty Acid Free BSA is ideally suited for fatty acid sensitive cell culture systems, such as CHO, Vero and MDCK cells, as well as for the most sensitive diagnostic and research applications
Catalog No. NC2112210 Supplier Gold Biotechnology Inc Supplier No. A4211
Add to Cart
Add to Cart
GoldBio's Reagent Grade, Fatty Acid Free BSA is ideally suited for fatty acid sensitive cell culture systems, such as CHO, Vero and MDCK cells, as well as for the most sensitive diagnostic and research applications