Learn More
Bioworld Bovine Serum Albumin (BSA), Fraction V - Molecular Biology Grade, 100 g
Supplier:  Bioworld 220700222
Encompass_Preferred
BSA fraction V, molecular biology grade is a highly purified, lyophilized powder. This product is processed to provide a minimum of 98% albumin and to be essentially free of both protease and nuclease activity.The pH is adjusted to 7.0. It is commonly used in PCR and other molecular biology applications.SpecificationsShips with cold packs
Catalog No. 50-253-932