Learn More
Bioworld Bovine Serum Albumin (BSA), Fraction V - Serum Replacement Grade, 20 g
Supplier:  Bioworld 220700201
Encompass_Preferred
Comparable to AlbuMAX, our serum-replacement grade BSA is a highly purified, lyophilized powder.Formulated to optimize the product’s lipid profile for growth promotion, this lipid-enriched BSA is processed to provide a minimum of 98% albumin and to be essentially free of IgG. This low-endotoxin preparation is intended for serum-free and serum-reduced cell culture applications. A supplementation rate ranging from 0.5 to 2.5 mg/mL has been effective in the production of recombinant proteins, virus particles and monoclonal antibodies in cultured cells.SpecificationsMycoplasma testedComplete 9 CFR 113.53c viral screenShips at ambient.
Catalog No. 50-253-925