Learn More
Medchemexpress LLC Bovine Serum Albumin, Carrier Protein | 9048-46-8 | ≥98% | 100 MG
SDP
Catalog No. 5000410224
Encompass_Preferred
Bovine Serum Albumin (BSA) is a 583 amino acid protein composed of three homologous full alpha structural domains. It is a spherical protein crucial for transporting molecules such as fatty acids, drugs, and hormones from the blood. BSA is utilized in various biochemical applications as a drug carrier for biologically active compounds. For long-term storage, recombinant protein solutions should be further diluted with 0.1% BSA.
  • Maintains protein stability and prevents protein hydrolysis and denaturation.
  • Acts as a blocking protein in immunoassays.
  • Serves as a cell culture medium supplement.
  • Used as standard protein in gel electrophoresis and protein assays.
  • Functions as a carrier protein.
Catalog No. 50-004-10224 Supplier Medchemexpress LLC Supplier No. HYCP002100MG
Bovine Serum Albumin (BSA) is a 583 amino acid protein composed of three homologous full alpha structural domains. It is a spherical protein crucial for transporting molecules such as fatty acids, drugs, and hormones from the blood. BSA is utilized in various biochemical applications as a drug carrier for biologically active compounds. For long-term storage, recombinant protein solutions should be further diluted with 0.1% BSA.
  • Maintains protein stability and prevents protein hydrolysis and denaturation.
  • Acts as a blocking protein in immunoassays.
  • Serves as a cell culture medium supplement.
  • Used as standard protein in gel electrophoresis and protein assays.
  • Functions as a carrier protein.