Learn More
Medchemexpress LLC Bovine Serum Albumin, Carrier Protein | 9048-46-8 | ≥98% | 50 MG
SDP
Catalog No. 5000411033
Encompass_Preferred
Bovine Serum Albumin (BSA) is a 583 amino acid protein composed of three homologous full alpha structural domains. BSA is a spherical protein crucial for transporting molecules like fatty acids, drugs, and hormones in the blood. It is utilized in various biochemical applications as a drug carrier for biologically active compounds. For long-term storage, recombinant protein solutions should be further diluted with 0.1% BSA.
  • Maintains protein stability and prevents protein hydrolysis and denaturation.
  • Blocks proteins in immunoassays.
  • Supplements cell culture medium.
  • Acts as a standard protein in gel electrophoresis and protein assays.
  • Functions as a carrier protein.
Catalog No. 50-004-11033 Supplier Medchemexpress LLC Supplier No. HYCP00250MG
Bovine Serum Albumin (BSA) is a 583 amino acid protein composed of three homologous full alpha structural domains. BSA is a spherical protein crucial for transporting molecules like fatty acids, drugs, and hormones in the blood. It is utilized in various biochemical applications as a drug carrier for biologically active compounds. For long-term storage, recombinant protein solutions should be further diluted with 0.1% BSA.
  • Maintains protein stability and prevents protein hydrolysis and denaturation.
  • Blocks proteins in immunoassays.
  • Supplements cell culture medium.
  • Acts as a standard protein in gel electrophoresis and protein assays.
  • Functions as a carrier protein.