Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the quantitative determination of Bovine Serum Albumin as a contaminant in various biological preparations. Assay Range: 156 - 10000 ng/ml; Sensitivity: 94 ng/ml.
Catalog No. NC2747093
Encompass_Preferred
Pack Size: 1 x 96 wells. This BSA ELISA can be used to optimize purification processes and in routine quality control of in-process streams, as well as in testing the final product. The kit employs a sandwich ELISAfor detecting Bovine Serum Albumin which leads to a higher specificity and increased sensitivity compared to conventional competitive ELISA kits which employ only one antibody. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Lot to Lot Consistency with Precision CV <10%.
Catalog No. NC2747093 Supplier KRISHGEN BIOSYSTEMS Supplier No. KBBP10
Pack Size: 1 x 96 wells. This BSA ELISA can be used to optimize purification processes and in routine quality control of in-process streams, as well as in testing the final product. The kit employs a sandwich ELISAfor detecting Bovine Serum Albumin which leads to a higher specificity and increased sensitivity compared to conventional competitive ELISA kits which employ only one antibody. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Lot to Lot Consistency with Precision CV <10%.