Learn More
Medchemexpress LLC Bovine Serum Albumin-FITC | 100 UL
SDP
Catalog No. 5000426157
Encompass_Preferred
Bovine Serum Albumin-FITC is bovine serum albumin conjugated to FITC (Ex=495 nm; Em=525 nm). It can be used as a fluorescent marker to label or detect specific antigens, molecules, or cellular structures.
  • Used as a fluorescent marker
  • Labels or detects specific antigens
  • Labels or detects specific molecules
  • Labels or detects specific cellular structures
Catalog No. 50-004-26157 Supplier Medchemexpress LLC Supplier No. HYNP051100UL
Bovine Serum Albumin-FITC is bovine serum albumin conjugated to FITC (Ex=495 nm; Em=525 nm). It can be used as a fluorescent marker to label or detect specific antigens, molecules, or cellular structures.
  • Used as a fluorescent marker
  • Labels or detects specific antigens
  • Labels or detects specific molecules
  • Labels or detects specific cellular structures