Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin Fraction V, 50 G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 10735078001
Encompass
From bovine serum. Bovine Serum Albumin Fraction V is suitable for standard applications such as: Stabilizer to prevent proteolysis and denaturation of enzymes and other proteins, especially when the proteins are present in low concentrations. Buffer component in immunochemistry, biochemistry, cell biology, or molecular biology, Reducing agent, Protein blocker for saturation of protein binding sites in ELISAs, Southern blots, and western blots, Media supplement for media that require addition of protein, Carrier protein, andStandard protein in gel electrophoresis and protein determination.
Catalog No. NC0954387