Learn More
Bioworld Bovine Serum Albumin (BSA), Heat Shock, Diagnostic Grade, 50 g
Supplier:  Bioworld 220700341
Encompass_Preferred
Specifications:Appearance: Lyophilized PowderColor: Off-white to tanMoisture: < 5.0%Ash: < 3.0%pH: 6.5 - 7.5Heavy Metals: < 20 ppmAlbumin: > 98.0%Protein: > 98%IgG: Not detectedProtease: Not detectedEndotoxin: < 3 EU/mgBioburden: Information onlyStorage: Store tightly sealed in a cool, dry environmentUsage: bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.
Catalog No. 50-253-958