Learn More
Bioworld Bovine Serum Albumin (BSA), Heat Shock, Protease-Free, 50 g
Supplier:  Bioworld 220700381
Encompass_Preferred
Prepared by a proprietary heat-shock process.Immunoglobulin content undetectable by our methods.Protease, DNASE, RNASE activity undetectable by our methods.Purity: > 98%.Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.
Catalog No. 50-253-963