Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin heat100G
Catalog No. 502907482
Encompass
Bovine serum albumin (BSA) a 66nbspkDa protein belongs to the family of serum albumins. It is an helical non-glycosylatednbspand globular protein nbspthat has 17-disulfidenbspbonds.
Catalog No. 50-290-7482 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A4919100G
Bovine serum albumin (BSA) a 66nbspkDa protein belongs to the family of serum albumins. It is an helical non-glycosylatednbspand globular protein nbspthat has 17-disulfidenbspbonds.